Z743363
Direct Thermal Cryo-Tags®
Cryo-Spot, blue, diam. Type="regular">1/2 in., pkg of 4000 roll of
Synonym(s):
cryogenic vial labels, freezer labels
About This Item
Recommended Products
packaging
pkg of 4000 roll of
manufacturer/tradename
Diversified Biotech DTCR-4500-B
parameter
temp. range at 70 to -196 °C
technique(s)
cryopreservation: suitable
diam.
Type="regular">1/2 in.
color
blue
compatibility
Important note for Customers outside of USA: Our Direct Thermal labels are designed to work with U.S. Model Dymo printers only. Labels are not compatible with European DYMO printers.
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Tough - Labels adhere to most plastics, glass and metals without cracking, peeling or degrading. Also resistant to scuffing and smearing
Compatible - Tubes labeled with Direct Thermal Cryo-Tags easily slide in and out of centrifuge rotors without binding
Specially Designed - Labels accept printing from direct thermal printers such as GoDEX DT200 Direct printers. Diversified Biotech recommends the Label LIVE app for printing in conjunction with the GoDEX DT200 printer.
Barcoding - Print popular barcode formats for fast and easy sample identification
Economical - Limit label waste by printing only the quantity required. Available in White, Blue, Green and Yellow
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service