Skip to Content
Merck
All Photos(1)

Documents

Z733423

IKA® ULTRA-TURRAX® disperser tools, plastic

dispersing element for T-10 basic disperser, 7 mm, 25/cs

Synonym(s):

disperser blades, tissue grinders, tissue homogenizers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

material

polycarbonate

feature

autoclavable
polycarbonate rotor-stator

packaging

case of 25
pack of 25

manufacturer/tradename

IKA 3433325

diam.

7 mm

Looking for similar products? Visit Product Comparison Guide

Other Notes

Shaft length for plastic dispersing element is 65 mm

Legal Information

IKA is a registered trademark of IKA-Werke GmbH & Co. KG
ULTRA-TURRAX is a registered trademark of IKA-Werke GmbH & Co. KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service