Key Documents
Z722839
Hettich® ROTOFIX 32A/Universal 320/320R centrifuge accessories
Rotor, 2 place for microplates, for UNIVERSAL 320
Select a Size
1 EA
CHF 764.00
CHF 764.00
Check Cart for Availability
Select a Size
Change View
1 EA
CHF 764.00
About This Item
UNSPSC Code:
41103900
NACRES:
NB.22
CHF 764.00
Check Cart for Availability
feature
to hold 2 x microtrite plates
packaging
pack of 1 ea
manufacturer/tradename
Hettich 1460
parameter
2,218 max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Universal 320/320R
Related Categories
General description
Carriers not included.
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. We are planning to get Hettich Universal 320R centrifuge with rotor for deep plate. After search website, I am not clear what is difference between Z722855 and Z722839? what is carriers that was not included in Z722855. Thanks
    1 answer
    1. Product Z722839 is a 2-place swing-out rotor for the Hettich Universal 320/320R. The manufacturer’s part number is 1460. This rotor has a maximum capacity of 10 plates.
      Product Z722855 is a 2-place swing-out rotor for the Hettich Rotina 380/380R. The manufacturer’s part number is 1760. This rotor has a maximum capacity of 10 plates.
      There is a slight difference in the maximum RCF, however these are not interchangeable. For the Universal 320/320R the Z722839 would be required.
      See the manufacturer’s images below:
      Helpful?