Skip to Content
Merck
All Photos(1)

Documents

Z710539

Hanna pH/temp meter

Model HI991002 pH/ORP/temp

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115603
NACRES:
NB.79

manufacturer/tradename

Hanna

Looking for similar products? Visit Product Comparison Guide

General description

The new HI 991001 and HI 991002 meters from Hanna offer IP 67 waterproof protection in a compact casing. The included multi-parameter electrode has a built-in temperature sensor. No additional temperature probe is required. The electrode also contains a mini amplifier to render your measurements impervious to noise and electrical interference. The large display shows readings in an extended range from -2.00 to 16.00 pH and simultaneously shows temperature from -5.0 to 105.0°C or 23 to 221°F. HI 991002 also measures ORP from -1999 to 1999 mV.

These meters have a stability indicator and hold feature that prompts you when to take the reading & freezes the display for easy and accurate recording. The large display also has graphic symbols to guide you through all operations. The battery life of the meters guarantee over 1500 hours of continuous use. At startup, they perform a self-check and then display the remaining battery level to assure proper working condition. Calibration is performed automatically at one or two points using standard or N.I.S.T. buffers.

Features and Benefits

  • Stability Indicator and Hold Feature insure proper readings
  • Battery Life of over 1500 Hours
  • Automatic Calibration using N.I.S.T

The new HI 991001 and HI 991002 meters from Hanna offer IP 67 waterproof protection in a compact casing. The included multi-parameter electrode has a built-in temperature sensor. No additional temperature probe is required. The electrode also contains a mini amplifier to render your measurements impervious to noise and electrical interference. The large display shows readings in an extended range from -2.00 to 16.00 pH and simultaneously shows temperature from -5.0 to 105.0°C or 23 to 221°F. HI 991002 also measures ORP from -1999 to 1999 mV.

The mV scale of this meter can be used with the available HI 1296D pH/temperature electrode to check the condition of the pH electrode. These meters have a stability indicator and hold feature that prompts you when to take the reading & freezes the display for easy and accurate recording. The large display also has graphic symbols to guide you through all operations. The battery life of the meters guarantee over 1500 hours of continuous use. At startup, they perform a self-check and then display the remaining battery level to assure proper working condition. Calibration is performed automatically at one or two points using standard or N.I.S.T. buffers.

HI 991001 is supplied complete with HI 1296D pH/temperature electrode, 3 × 1.5 V AA batteries and carrying case.
HI 991002 is supplied complete with HI 1297D pH/ORP/temperature electrode, 3 × 1.5 V AA batteries and carrying case.

SpecificationsHI 991001HI 991002

Ranges:pH-2.00 to 16.00 -2.00 to 16.00
ORP-±1999 mV
Temperature-5.0 to 105.0°C or 23.0 to 221.0°F-5.0 to 105.0°C or 23.0 to 221.0°F

Resolution:
pH0.010.01
ORP-1 mV
Temperature0.1°C or 0.1°F0.1°C or 0.1°F

Accuracy:
pH±0.02±0.02
(@20°C/68°F):ORP-±2 mV
Temperature±0.5 up to 60°C, ±1°C up to 105°C±0.5 up to 60°C, ±1°C up to 105°C
±1°F up to 140°F, ±2°F up to 221°F±1°F up to 140°F, ±2°F up to 221°F
Typical EMC pH±0.02±0.02
Deviation:ORP-±2 mV
Temperature±0.2°C or ±0.4°F±0.2°C or ±0.4°F

pH Calibration:
Automatic 1 or 2 points with 2 set of standardized buffers
(4.01, 7.01, 10.01 or 4.01, 6.86, 9.18)

Temp. Compensation:
Automatic from -5 to 105°C (23 to 221°F) (for pH only)

Electrode:
HI 1296D combination amplified pH/temperature HI 1297D combination amplified pH/ORP/temperature
with DIN connector and 1 m (3.3′) cable (included) with DIN connector and 1 m (3.3′) cable (included)

Environment:
0 to 50°C (32 to 122°F); RH 100%
Battery Type:3 × 1.5 V AA
Battery Life:Approximately 1500 hours of continuous use. Auto-off after 8 minutes
Dimensions:150 × 80 × 36 mm (5.9 × 3.1 × 1.4")
Weight:245 g (8.6 oz.)

Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance

Components

HI 991002 is supplied complete with HI 1296D pH/temperature electrode, 3 × 1.5 V AA batteries and carrying case.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service