Z692859
microDOC Compact Gel Documentation System, with UV Transilluminator (UVT254/365)
with TotalLab™ 1D software, AC/DC input 230 V AC
All Photos(2)
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
feature
Contains UK and Schuko plugs
manufacturer/tradename
Cleaver CSI-MDOCUV254/3651D
General description
System comprises of:
Canon G12 digital
2MB storage card
Ethidium bromide & close up +3 filters
Lift-off hood with easy access door and built-in inner lights
8" TFT colour monitor
12 x 12 cm UV 254/365 nm dual tranilluminator
Canon G12 digital
2MB storage card
Ethidium bromide & close up +3 filters
Lift-off hood with easy access door and built-in inner lights
8" TFT colour monitor
12 x 12 cm UV 254/365 nm dual tranilluminator
This option combines the microDOC Compact Gel Documentation System with a 12 x 12 cm UV 254/365 nm dual tranilluminator.
Available with optional TotalLab™ 1D software. Main software functions include: lane creation, background subtraction, band detection, profile deconvolution, and molecular size, pI and quantity calibration.
Available with optional TotalLab™ 1D software. Main software functions include: lane creation, background subtraction, band detection, profile deconvolution, and molecular size, pI and quantity calibration.
Legal Information
TotalLab is a trademark of Nonlinear Dynamics Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)