Key Documents
Z659630
Bandelin SONOREX Ultrasonic bath accessories
insert tub with lid for Z659584, Z659592 and Z659002
Select a Size
1 EA
CHF 86.60
CHF 86.60
Estimated to ship on25 May 2025
Select a Size
Change View
1 EA
CHF 86.60
About This Item
UNSPSC Code:
41103706
NACRES:
NB.74
CHF 86.60
Estimated to ship on25 May 2025
material
polyethylene
packaging
pack of 1 ea
manufacturer/tradename
Bandelin KW 3
availability
not available in USA and Canada
L × W × H
195 mm × 115 mm × 88 mm
General description
Stainless steel lid for an ultrasonic bath protects the bath liquid from external contamination and prevents unpleasant odors from the cleaning or disinfection agents added. In addition, the noise emitted when the ultrasonic cleaner is in operation, which may be perceived as annoying, is reduced. The lids are designed in such a way that any condensation that occurs is directed back into the bath.
Features and Benefits
  • protects the bath fluid from contamination
  • sound-absorbing
Legal Information
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)