Skip to Content
Merck
All Photos(1)

Documents

Z601179

Hettich® MIKRO 220/220R centrifuge accessories

for MIKRO 220R Centrifuge, Adapter for 50 mL Falcon tubes (29 x 115 mm)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103911
NACRES:
NB.22

feature

Holds 50 mL tubes

packaging

pack of 3 ea

manufacturer/tradename

Hettich 1641

technique(s)

centrifugation: suitable

compatibility

for use with 1016, 1137 (rotors)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

  • compatible with rotor 1016 (Z601381)
  • rotor 1016 (Z601381) is only compatible with the MIKRO 220R Centrifuge

Legal Information

Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service