Skip to Content
Merck
All Photos(2)

Key Documents

X2629

Sigma-Aldrich

endo-1,4-β-Xylanase from Trichoderma longibrachiatum

greener alternative

Synonym(s):

1,4-β-D-Xylanxylanohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

form

powder

Quality Level

specific activity

≥1.0 units/mg solid

composition

Protein, ≥10%

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

greener alternative category

storage temp.

room temp

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for waste prevention when used in cellulosic ethanol research. For more information see the article in biofiles.

Biochem/physiol Actions

Primary activity is an acid-neutral endo-1,4-β-D-xylanase, additional activities include β-glucanase, cellulase, pectinase, mannanase, xyloglucanase, laminarase, β-glucosidase, β-xylosidase, α-L-arabinofuranosidase, amylase, and protease.

Unit Definition

One unit will liberate 1 μmole of reducing sugar measured as xylose equivalents from xylan (Cat. No. X0627) per min at pH 4.5 at 30 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Miao et al.
Frontiers in bioengineering and biotechnology, 9, 690702-690702 (2021-09-14)
Xylanases with high specific activity has been focused with great interest as a useful enzyme in biomass utilization. The production of recombinant GH11 xylanase (MYCTH_56237) from Myceliophthora thermophila has been improved through N-terminal signal peptide engineering in P. pastoris. The
Jiao Tang et al.
Applied microbiology and biotechnology, 103(7), 3037-3048 (2019-02-15)
The thermophilic fungus Thielavia terrestris when cultured on cellulose produces a cocktail of thermal hydrolases with potential application in saccharification of lignocellulosic biomass and other biotechnological areas. Glucuronoyl esterases are considered to play a unique role as accessory enzymes in
Zhenghui Liu et al.
Frontiers in bioengineering and biotechnology, 9, 814246-814246 (2022-02-15)
Supplementing commercial xylanase and cellulase with selected debranching enzymes only resulted in slight enhancement of the enzymatic hydrolysis of wheat bran autohydrolysis residues (WBAR) which was obtained at 160°C over a 30-min period of autohdyrolysis, while a blend of enzymes
Padil et al.
RSC advances, 13(31), 21403-21413 (2023-07-19)
Tetraselmis chuii is a potential microalgae that is in consideration for producing bioethanol owing to its large content of carbohydrates. The glucose production from T. chuii through an enzymatic process with cellulase and xylanase (pretreatment process) and α-amylase and glucoamylase
Dandan Li et al.
Biotechnology for biofuels, 13, 106-106 (2020-06-17)
The sorghum stem can be divided into the pith and rind parts with obvious differences in cell type and chemical composition, thus arising the different recalcitrance to enzyme hydrolysis and demand for different pretreatment conditions. The introduction of organic solvents

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service