Skip to Content
Merck
All Photos(4)

Key Documents

WH0055145M1

Sigma-Aldrich

Monoclonal Anti-THAP1 antibody produced in mouse

clone 2C1-2F2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-FLJ10477, Anti-MGC33014, Anti-THAP domain containing, apoptosis associated protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 474.00

CHF 474.00


Check Cart for Availability


Select a Size

Change View
100 μG
CHF 474.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 474.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2C1-2F2, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... THAP1(55145)

General description

THAP domain containing 1 (THAP1) is a transcription factor encoded by the gene mapped to human chromosome 18p11.21. The encoded protein contains an N-terminal conserved DNA-binding domain, designated THAP domain with a zinc-finger structure.

Immunogen

THAP1 (AAH21721, 1 a.a. ~ 213 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MVQSCSAYGCKNRYDKDKPVSFHKFPLTRPSLCKEWEAAVRRKNFKPTKYSSICSEHFTPDCFKRECNNKLLKENAVPTIFLCTEPHDKKEDLLEPQEQLPPPPLPPPVSQVDAAIGLLMPPLQTPVNLSVFCDHNYTVEDTMHQRKRIHQLEQQVEKLRKKLKTAQQRCRRQERQLEKLKEVVHFQKEKDDVSERGYVILPNDYFEIVEVPA

Biochem/physiol Actions

THAP domain containing 1 (THAP1) plays a key role in endothelial cell proliferation and pro-apoptotic processes. Mutation in the gene is associated with primary dystonia type 6 (DYT6). The encoded protein links prostate-apoptosis-response-4 (Par-4) to promyelocytic leukemia (PML) nuclear bodies, which facilitates both serum withdrawal and tumor necrosis factor-α (TNF-α)-induced apoptosis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intrafamilial variability of the primary dystonia DYT6 phenotype caused by p.Cys5Trp mutation in THAP1 gene.
Jurek M, et.al
Neurologia i Neurochirurgia Polska, 48(4), 254-257 (2014)
Interaction between variants of two glycosyltransferase genes in IgA nephropathy.
Zhu L, et.al
Kidney International, 76(2), 190-198 (2009)
Screening for THAP1 Mutations in Polish Patients with Dystonia Shows Known and Novel Substitutions.
Golanska E
PLoS ONE, 10(6), e0129656-e0129656 (2015)
Marta Jurek et al.
Neurologia i neurochirurgia polska, 48(4), 254-257 (2014-08-30)
Mutations localized in THAP1 gene, locus 18p11.21 have been reported as causative of primary dystonia type 6 (DYT6). Disease which is characterized mainly by focal dystonia, frequently involving the craniocervical region, however associated also with early-onset generalized dystonia and spasmodic
Li Zhu et al.
Kidney international, 76(2), 190-198 (2009-04-10)
Increasing evidence points to the importance of aberrant O-glycosylated immunoglobulin A1 (IgA1) in the pathogenesis of IgA nephropathy (IgAN), a disease widely considered to be a polygenic disorder. We earlier found that haplotypes in two key glycosyltransferase genes, C1GALT1 and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service