Skip to Content
Merck
All Photos(2)

Key Documents

WH0010660M1

Sigma-Aldrich

Monoclonal Anti-LBX1 antibody produced in mouse

clone 2B2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-HPX6, Anti-LBX1H, Anti-homeobox, Anti-ladybird homeobox homolog 1 (Drosophila)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 474.00

CHF 474.00


Check Cart for Availability


Select a Size

Change View
100 μG
CHF 474.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 474.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2B2, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LBX1(10660)

General description

This gene and the orthologous mouse gene were found by their homology to the Drosophila lady bird early and late homeobox genes. In the mouse, this gene is a key regulator of muscle precursor cell migration and is required for the acquisition of dorsal identities of forelimb muscles. (provided by RefSeq)

Immunogen

LBX1 (NP_006553.2, 133 a.a. ~ 219 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
TNHQIYELEKRFLYQKYLSPADRDQIAQQLGLTNAQVITWFQNRRAKLKRDLEEMKADVESAKKLGPSGQMDIVALAELEQNSEATA

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service