Skip to Content
Merck
All Photos(2)

Key Documents

WH0002262M1

Sigma-Aldrich

Monoclonal Anti-GPC5 antibody produced in mouse

clone 1C9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-glypican 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C9, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPC5(2262)

General description

Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation. (provided by RefSeq)

Immunogen

GPC5 (AAH39730, 1 a.a. ~ 572 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MDAQTWPVGFRCLLLLALVGSARSEGVQTCEEVRKLFQWRLLGAVRGLPDSPRAGPDLQVCISKKPTCCTRKMEERYQIAARQDMQQFLQTSSSTLKFLISRNAAAFQETLETLIKQAENYTSILFCSTYRNMALEAAASVQEFFTDVGLYLFGADVNPEEFVNRFFDSLFPLVYNHLINPGVTDSSLEYSECIRMARRDVSPFGNIPQRVMGQMGRSLLPSRTFLQALNLGIEVINTTDYLHFSKECSRALLKMQYCPHCQGLALTKPCMGYCLNVMRGCLAHMAELNPHWHAYIRSLEELSDAMHGTYDIGHVLLNFHLLVNDAVLQAHLNGQKLLEQVNRICGRPVRTPTQSPRCSFDQSKEKHGMKTTTRNSEETLANRRKEFINSLRLYRSFYGGLADQLCANELAAADGLPCWNGEDIVKSYTQRVVGNGIKAQSGNPEVKVKGIDPVINQIIDKLKHVVQLLQGRSPKPDKWELLQLGSGGGMVEQVSGDCDDEDGCGGSGSGEVKRTLKITDWMPDDMNFSDVKQIHQTDTGSTLDTTGAGCAVATESMTFTLISVVMLLPGIW

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service