Skip to Content
Merck
All Photos(4)

Key Documents

WH0000817M2

Sigma-Aldrich

Monoclonal Anti-CAMK2D antibody produced in mouse

clone 1A8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CAMKD, Anti-MGC44911, Anti-calcium/calmodulin-dependent protein kinase (CaM kinase) II delta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A8, monoclonal

form

buffered aqueous solution

species reactivity

mouse

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2bλ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAMK2D(817)

General description

The gene encoding calcium/calmodulin dependent protein kinase IIδ (CAMK2D) is localized on human chromosome 4q26. The encoded protein is a member of serine/threonine protein kinase family and the Ca2+/calmodulin-dependent protein kinase subfamily.
The product of this gene belongs to the serine/threonine protein kinase family and to the Ca2+/calmodulin-dependent protein kinase subfamily. Calcium signaling is crucial for several aspects of plasticity at glutamatergic synapses. In mammalian cells, the enzyme is composed of four different chains: α, β, γ, and δ. The product of this gene is a δ chain. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Distinct isoforms of this chain have different expression patterns

Immunogen

CAMK2D (AAH32784, 301 a.a. ~ 410 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
KGAILTTMLATRNFSAAKSLLKKPDGVKESTESSNTTIEDEDVKARKQEIIKVTEQLIEAINNGDFEAYTKICDPGLTAFEPEALGNLVEGMDFHRFYFENALSKSNKPI

Biochem/physiol Actions

As a subunit of the calcium/calmodulin dependent protein kinase II complex, calcium/calmodulin dependent protein kinase II complex δ (CAMK2D) modulates the functions of ion channels and Ca2+ handling proteins. It also regulates the phosphorylation and acetylation of histones. CAMK2D has been shown to be overexpressed in the myocardium of diabetic patients.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Control of histone H3 phosphorylation by CaMKII? in response to haemodynamic cardiac stress.
Awad S
The Journal of Pathology, 235, 606-618 (2015)
Genes for Fibrogenesis in the Determination of Susceptibility to Myocardial Infarction
Goncharova IA
Molekuliarnaia Biologiia, 50, 94-105 (2016)
Inhibition of elevated Ca2+/calmodulin-dependent protein kinase II improves contractility in human failing myocardium.
Sossalla S
Circulation Research, 107, 1150-1161 (2010)
The role of CaMKII in diabetic heart dysfunction.
Daniels L
Heart Failure Reviews, 20, 589-600 (2015)
Joshua Abd Alla et al.
Nature cardiovascular research, 2(11), 1044-1059 (2023-01-01)
Tetralogy of Fallot (TOF) is one of the most prevalent congenital heart defects, with adverse cardiac remodeling and long-term cardiac complications. Here, searching for pathomechanisms, we find upregulated bublin coiled-coil protein (BBLN) in heart specimens of TOF patients with cyanosis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service