Skip to Content
Merck
All Photos(1)

Key Documents

V5764

Sigma-Aldrich

Anti-Vesicular GABA Transporter antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-VGAT

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.77

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

potency

2-4 mg per mL effective concentration (determined by immunoblotting, using mouse brain and rat brain extracts)

mol wt

antigen ~55 kDa

species reactivity

rat, mouse

packaging

pkg of 200 μL

storage condition

avoid repeated freeze/thaw cycles (Storage in "frost-free" freezers is not recommended)

concentration

~1.0 mg/mL

technique(s)

microarray: suitable
western blot: 2-4 μg/mL using mouse brain and rat brain extracts (S1 fraction)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Vesicular GABA transporter (VGAT), also known as solute carrier family 32 member 1 (SLC32A1) or vesicular inhibitory amino acid transporter (VIAAT), is encoded by the gene localized on human chromosome 20q11.23. The encoded protein is highly expressed in both γ-aminobutyric acid -ergic (GABAergic) and glycinergic nerve endings.

Immunogen

synthetic peptide corresponding to amino acids 1-20 located at the N-terminus of mouse VGAT. This sequence is identical in rat VGAT and highly conserved (single a.a. substitution) in human VGAT.

Application

Anti-Vesicular GABA Transporter antibody produced in rabbit has been used in:
  • immunohistochemistry
  • western blot
  • immunocytochemistry

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Vesicular GABA transporter (VGAT) plays a vital role in loading of presynaptic vesicles with glycine and γ-aminobutyric acid (GABA). VGAT is precisely involved in packing GABA into synaptic vesicles. In addition, it also has a crucial role in inhibitory synaptic transmission. Genetic variations in the gene leads to hyperekplexia.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Distribution and Posttranslational Modification of Synaptic ERα in the Adult Female Rat Hippocampus
Tabatadze N
Endocrinology, 154, 2013-2013 (2013)
Forebrain Ischemia Triggers GABAergic System Degeneration in Substantia Nigra at Chronic Stages in Rats
Lin B
Cardiovascular Psychiatry and Neurology (2010)
Cyclin-Dependent Kinase 5 Is Involved in the Phosphorylation of Gephyrin and Clustering of GABAA Receptors at Inhibitory Synapses of Hippocampal Neurons
Kalbouneh H
PLoS ONE, 9 (2014)
B Lin et al.
Cardiovascular psychiatry and neurology, 2010, 506952-506952 (2010-10-29)
The long-term consequences of forebrain ischemia include delayed Parkinson's syndrome. This study revealed delayed neurodegeneration in the substantia nigra 8 weeks after 12.5 minutes of global ischemia in rat brain. Following neuronal loss of 30-40% in central and dorsolateral striatum
A critical role for glycine transporters in hyperexcitability disorders
Harvey RJ
Frontiers in Molecular Neuroscience (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service