Skip to Content
Merck
All Photos(3)

Documents

U5385

Sigma-Aldrich

UNISETTE Biopsy Processing/Embedding Cassettes

blue

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124
NACRES:
NA.47

color

blue

application(s)

hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Histology processing/embedding cassettes made of acetal polymer. These cassettes keep specimens safely submerged in liquid and are resistant to histological solvents. The one-piece lid snaps apart and conveniently locks into the cassette base, ensuring specimen safety. A line of cassette colors are available to fit your needs and preference. A large 30° anterior writing area is available for your convenience. Not suitable for automated printers.

Application

Compatible with Leica and Sakura printers.

Quantity

Case contains 3 dispenser boxes of 500 cassettes.

Legal Information

Unisette is a trademark of Simport

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service