Skip to Content
Merck
All Photos(1)

Key Documents

TMO312114-0016

Nalgene® wide-mouth square bottle

high-density polyethylene, capacity 500 mL, natural, case of 125 ea

Synonym(s):

clear plastic bottle, translucent bottle

Sign Into View Organizational & Contract Pricing

Select a Size

125 EACH
CHF 726.00

CHF 726.00


Estimated to ship on16 April 2025



Select a Size

Change View
125 EACH
CHF 726.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

CHF 726.00


Estimated to ship on16 April 2025


material

high-density polyethylene
square
wide-mouth

packaging

case of 125 ea

manufacturer/tradename

Nalgene 312114-0016

capacity

500 mL

color

natural

Looking for similar products? Visit Product Comparison Guide

General description

Easily fill and pour with Thermo Scientific Nalgene Square Wide-Mouth HDPE Bottles with Closure, featuring molded-in graduations for easy measuring and durable, uniform walls that are resistant to splitting or puncturing. These translucent bottles provide visible liquid levels and come in a space-saving square design.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service