Skip to Content
Merck
All Photos(1)

Key Documents

T8828

Sigma-Aldrich

Monoclonal Anti-TIGAR antibody produced in mouse

~1.0 mg/mL, clone 9C10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Monoclonal Anti-C12ORF5, Monoclonal Anti-Chromosome 12 open reading frame 5, Monoclonal Anti-TP53-Induced glycolysis and apoptosis regulator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9C10, monoclonal

form

buffered aqueous solution

mol wt

antigen ~30 kDa

species reactivity

monkey, human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 2-4 μg/mL using HeLa nuclear extract

isotype

IgG2b

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... C12orf5(57103)

General description

Monoclonal Anti-TIGAR (mouse IgG2b isotype) is derived from the hybridoma 9C10 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a peptide corresponding to a fragment in exon 6-encoding region of human TIGAR . TIGAR expression was found to correlate with fludarabine sensitivity in chronic lymphocytic leukemia (CLL) cells.
TP53-induced glycolysis and apoptosis regulator (TIGAR) is a p53 target gene mapped to human chromosome 12p13-3. The gene contains six exons and it is characterized with two possible p53 binding sites, one upstream of the first exon (BS1) and one within the first intron (BS2).

Application

Monoclonal Anti-TIGAR antibody produced in mouse has been used in:
  • enzyme linked immuno sorbent assay (ELISA)
  • immunoblotting
  • immunoprecipitation

Biochem/physiol Actions

TP53-induced glycolysis and apoptosis regulator (TIGAR) inhibits glycolysis by decreasing the fructose-2, 6-bisphosphate levels in cells. This p53 induced protein also plays a vital role in regulation of intracellular reactive oxygen species (ROS) levels. TIGAR expression is associated with oncogenesis and development of several human cancers. Overexpression of TIGAR is observed in colorectal cancer patients. Thus, TIGAR expression might function as a potential biomarker for diagnosis of colorectal cancer and it can act as a target for developing therapeutics for the treatment of colorectal cancer.[1] Nullified expression of TIGAR, increases the radiosensitivity of tumor cells and also enhances the autophagy activity in cells with nutrient starvation or metabolic stress. TIGAR increases cancer cell survival rate in response to tumor chemotherapy by inhibiting both apoptosis and autophagy.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khayal Al-Khayal et al.
Oncology reports, 35(3), 1281-1286 (2015-12-18)
The TP53-induced glycolysis and apoptosis regulator (TIGAR) is a p53 target gene known to regulate glycolysis by acting as fructose bis-phosphatase (FBPase) and modulate reactive oxygen species. TIGAR expression has been implicated in oncogenesis and progression of several human cancers.
Guomei Tai et al.
International journal of clinical and experimental pathology, 8(5), 4823-4829 (2015-07-21)
Our previous study proved that TP53-induced glycolysis and apoptosis regulator (TIGAR) abrogation is able to radiosensitize glioma cells. Whether TIGAR over-expression has radio-protective effect in human parotid gland cells is still unknown. In this study human parotid gland fibroblast Hs
Z-B Hu et al.
Cancer gene therapy, 15(3), 173-182 (2007-12-25)
Oncolytic adenoviruses, also called conditionally replicating adenoviruses (CRADs), have been widely applied in cancer gene therapy. However, the construction of CRADs is still time-consuming. In this study, we attempted to establish a simplified method of generating CRADs based on AdEasy
Karim Bensaad et al.
Cell, 126(1), 107-120 (2006-07-15)
The p53 tumor-suppressor protein prevents cancer development through various mechanisms, including the induction of cell-cycle arrest, apoptosis, and the maintenance of genome stability. We have identified a p53-inducible gene named TIGAR (TP53-induced glycolysis and apoptosis regulator). TIGAR expression lowered fructose-2,6-bisphosphate
Mónica López-Guerra et al.
Haematologica, 93(12), 1843-1851 (2008-10-24)
The nucleoside analogue fludarabine is used in the treatment of chronic lymphocytic leukemia. It triggers p53-mediated apoptosis, although the mutational status of p53 does not fully account for heterogeneity in responsiveness to treatment. The aim of this study was to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service