Skip to Content
Merck
All Photos(1)

Key Documents

T8455

Sigma-Aldrich

Tyrosinase Fragment 369-377 human

Synonym(s):

CMM8, OCA1A, OCAIA, SHEP3, TYR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H66N10O16S2
CAS Number:
Molecular Weight:
1031.16
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

Quality Level

Assay

97-100% (HPLC)

form

lyophilized powder

color

white

solubility

deionized water: soluble

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C42H66N10O16S2/c1-20(2)33(42(67)68)52-39(64)25(10-11-30(44)56)47-40(65)29(19-53)50-38(63)27(13-15-70-5)48-41(66)34(21(3)54)51-31(57)18-45-36(61)28(17-32(58)59)49-37(62)26(12-14-69-4)46-35(60)24(43)16-22-6-8-23(55)9-7-22/h6-9,20-21,24-29,33-34,53-55H,10-19,43H2,1-5H3,(H2,44,56)(H,45,61)(H,46,60)(H,47,65)(H,48,66)(H,49,62)(H,50,63)(H,51,57)(H,52,64)(H,58,59)(H,67,68)

InChI key

MFDQNJLLKIATDB-UHFFFAOYSA-N

Gene Information

human ... TYR(7299)

Amino Acid Sequence

Tyr-Met-Asp-Gly-Thr-Met-Ser-Gln-Val-TFA

Application

Tyrosinase Fragment 369-377 human is a peptide used for controlling the melanin production.

Biochem/physiol Actions

The Tyrosinase enzyme catalyzes the first steps in the conversion of tyrosine to melanin. This is a membrane bound enzyme expressed in primary and metastatic melanoma lesions. The Tyrosine fragment 369-377 peptide is presented by HLA-A2 antigen to CTLs.
Tyrosinase Fragment 369-377 human is a peptide fragment of enzyme tyrosinase belongs to tyrosinase family. It catalyzes three distinct reactions, the hydroxylation of a monophenol, the dehydrogenation of a catechol and the dehydrogenation of a dihydroxyindole during biosynthesis of melanin.[1]

Physical form

Lyophilized from 0.1% aqueous TFA

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Körner et al.
Science (New York, N.Y.), 217(4565), 1163-1165 (1982-09-17)
The biosynthesis of melanin is initiated by the catalytic oxidation of tyrosine to dopa by tyrosinase in a reaction that requires dopa as a cofactor. Tyrosine then catalyzes the dehydrogenation of dopa to dopaquinone. The subsequent reactions can proceed spontaneously

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service