Skip to Content
Merck
All Photos(4)

Key Documents

T8076

Sigma-Aldrich

Anti-TRB3 (N-terminal) antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Human tribbles 3 homolog, Anti-NIPK, Anti-SINK, Anti-SKIP3, Anti-TRIB3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 39 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

western blot: 1-2 μg/mL using HEK-293 cells expressing human TRB3

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRIB3(57761)

General description

Tribbles homolog 3 (TRB3) gene is located on the 20p13 human chromosome. TRB3 and its related family members TRB1 and TRB2 show 45% sequence identity. It exhibits resemblance to ′tribbles′, a drosophila protein. TRB family members have a truncated kinase domain that lacks an adenosine 5′-triphosphate binding site (GXGXXG) and contain a variant catalytic core motif.

Immunogen

synthetic peptide corresponding to amino acids 11-29 located near the N-terminus region of human TRB3. The immunizing sequence has limited homology (~50% identity) to mouse and rat TRB3, and has no identity to TRB1 and TRB2 isoforms.

Application

Anti-TRB3 (N-terminal) antibody produced in rabbit has been used in immunoprecipitation and immunoblotting.

Biochem/physiol Actions

Tribbles homolog 3 (TRB3) like tribbles has also been identified as a potential pro-apoptotic protein, and is induced by a variety of cell stress conditions, including hypoxia and nutrient starvation. It was previously identified as a neuronal cell death-inducible protein kinase (NIPK) that is rapidly upregulated during neuronal apoptosis. TRB3 binds to activating transcription factor 4 (ATF4) and inhibits its transcriptional activity. TRB3 has been shown to interact with mitogen-activated protein kinases (MAPK) kinases to regulate activation of MAPK pathways.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Smyd1 facilitates heart development by antagonizing oxidative and ER stress responses
Rasmussen TL, et al.
PLoS ONE, 10(3), e0121765-e0121765 (2015)
CHOP10/TRB3/Akt signaling regulates ER stress apoptosis in colon cancer cells treated with prostamide-J
Albassam H, et al.
American Journal of Physiology: Renal Physiology (2019)
TRB3: an oxidant stress-induced pseudokinase with a potential to negatively modulate MCP-1 cytokine in diabetic nephropathy
Kanwar YS
American Journal of Physiology: Renal Physiology, 299(5), F963-F964 (2010)
Downregulation of TRB3 protects neurons against apoptosis induced by global cerebral ischemia and reperfusion injury in rats
Wei K, et al.
Neuroscience, 360, 118-127 (2017)
Role of TRIB3 in diabetic and overnutrition-induced atherosclerosis
Sowers JR
Diabetes, 61(2), 265-266 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service