Skip to Content
Merck
All Photos(1)

Key Documents

T7330

Sigma-Aldrich

Teijin compound 1 hydrochloride

≥98% (HPLC), powder

Synonym(s):

N-[2-[[(3R)-1-[(4-chlorophenyl)methyl]-3-pyrrolidinyl]amino]-2-oxoethyl]-3-(trifluoromethyl)benzamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21ClF3N3O2·HCl
CAS Number:
Molecular Weight:
476.32
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

Cl.FC(F)(F)c1cccc(c1)C(=O)NCC(=O)N[C@@H]2CCN(C2)Cc3ccc(Cl)cc3

InChI

1S/C21H21ClF3N3O2.ClH/c22-17-6-4-14(5-7-17)12-28-9-8-18(13-28)27-19(29)11-26-20(30)15-2-1-3-16(10-15)21(23,24)25;/h1-7,10,18H,8-9,11-13H2,(H,26,30)(H,27,29);1H/t18-;/m1./s1

InChI key

PGUQBBISBKGQDC-GMUIIQOCSA-N

Application

Studies have reported that Trp982.60, Thr2927.40, His1213.33 and Ile2636.55 of Teijin compound 1 significantly affect the antagonistic functions of the compound at CCR2[1].

Biochem/physiol Actions

Teijin compound 1 is CCR2b receptor antagonist. Teijin compound 1 inhibits cell chemotaxis induced by MCP-1. The chemokine receptors CCR2 is G protein-coupled receptors (GPCR), and is mainly expressed in monocytes, immature dendritic cells, activated T-lymphocytes and basophils.

Preparation Note

Teijin compound 1 hydrochloride is soluble in DMSO at a concentration that is greater than or equal to 20 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spencer E Hall et al.
Molecular pharmacology, 75(6), 1325-1336 (2009-03-20)
Design of dual antagonists for the chemokine receptors CCR2 and CCR5 will be greatly facilitated by knowledge of the structural differences of their binding sites. Thus, we computationally predicted the binding site of the dual CCR2/CCR5 antagonist N-dimethyl-N-[4-[[[2-(4-methylphenyl)-6,7-dihydro-5H-benzohepten-8-yl] carbonyl]amino]benzyl]tetrahydro-2H-pyran-4-aminium (TAK-779)

Articles

We offer many products related to chemokine receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service