Skip to Content
Merck
All Photos(1)

Key Documents

T2824

Sigma-Aldrich

Monoclonal Anti-Tumor Necrosis Factor-α antibody produced in hamster

~2 mg/mL, clone TN3-19.12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-TNF-α

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
CHF 157.00
200 μL
CHF 544.00

CHF 157.00


Check Cart for Availability


Select a Size

Change View
25 μL
CHF 157.00
200 μL
CHF 544.00

About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

CHF 157.00


Check Cart for Availability

biological source

hamster

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

TN3-19.12, monoclonal

form

buffered aqueous solution

species reactivity

rat, mouse

packaging

antibody small pack of 25 μL

concentration

~2 mg/mL

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
microarray: suitable
neutralization: suitable
western blot: 0.25-0.5 μg/mL using recombinant mouse TNF-α

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Tnf(21926)
rat ... Tnf(24835)

General description

Monoclonal Anti-Mouse Tumor Necrosis Factor-α (TNF-α) is derived from the TN3-19.12 hybridoma produced by the fusion of mouse myeloma cells (P3X63-Ag8.653) and splenocytes from male Armenian hamster immunized with purified mouse recombinant TNF-α. Tumor necrosis factor (TNF) protein family contains two members: TNF-α (also known as cachectin) and TNF-β (also known as lymphotoxin). These proteins are small molecules with a molecular weight of 17 and 25 kDa respectively and share 30% amino acid homology. TNF-α and -β are secreted from many different cells such as lymphocytes, neutrophils, and macrophages.

Specificity

Mouse monoclonal clone TN3-19.12 anti-Tumor Necrosis Factor-α antibody recognizes mouse and rat TNF-α (approx. 17 kDa in its monomer form and 51 kDa in its homotrimer form) and cross-reacts with mouse TNF-β.

Immunogen

purified recombinant mouse TNF-α.

Application

Monoclonal Anti-Tumor Necrosis Factor-α antibody produced in hamster has been used in:
  • immunoblotting
  • enzyme-linked immunosorbent assay (ELISA)
  • immunoprecipitation
  • neutralization

Biochem/physiol Actions

TNF-α is a major mediator of inflammatory responses and its over-production results in extensive tissue damage associated with endotoxemia and cachexia. High serum levels of TNF are associated with septic shock, rejection of renal transplants, parasitic infections, and various neoplastic diseases. TNF-α exhibits growth-enhancing activities for normal cells like fibroblasts, T and B cells, activation of endothelial cells, and differentiation of myeloid cells. In neoplastic cells, TNF-α exerts growth inhibitory activities that can be either cytostatic or cytotoxic.

Physical form

Supplied as a 0.2 μm filtered solution in 0.01 M phosphate buffered saline, pH 7.4.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of the Adipocyte in Metabolism and Endocrine Function
Endocrinology: Adult and Pediatric (2016)
Cytokines, inflammation and pain
Zhang JM and An J
International Anesthesiology Clinics, 45(2), 27-27 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service