Skip to Content
Merck
All Photos(2)

Key Documents

T0325

Sigma-Aldrich

Anti-TRPC5 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-transient receptor potential cation channel, subfamily C, member 5

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
CHF 328.00
200 μL
CHF 833.00

CHF 328.00


Estimated to ship on25 May 2025



Select a Size

Change View
50 μL
CHF 328.00
200 μL
CHF 833.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

CHF 328.00


Estimated to ship on25 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

western blot: 1:200 using rat brain membranes

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRPC5(7224)
mouse ... Trpc5(22067)
rat ... Trpc5(140933)

Immunogen

synthetic peptide corresponding to amino acid residues 959-973 of human TRPC5. This sequence is identical in rat, mouse, and rabbit.

Physical form

Lyophiized from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chase M Carver et al.
Frontiers in neuroscience, 15, 681144-681144 (2021-09-08)
We investigated the contribution of excitatory transient receptor potential canonical (TRPC) cation channels to posttraumatic hyperexcitability in the brain 7 days following controlled cortical impact model of traumatic brain injury (TBI) to the parietal cortex in male adult mice. We
M Salomonsson et al.
Acta physiologica (Oxford, England), 200(3), 265-278 (2010-04-30)
In rat afferent arterioles we investigated the role of Na(+) entry in noradrenaline (NA)-induced depolarization and voltage-dependent Ca(2+) entry together with the importance of the transient receptor potential channel (TRPC) subfamily for non-voltage-dependent Ca(2+) entry. R (340/380) Fura-2 fluorescence was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service