T0280
Tetramethylammonium hydroxide solution
25 % (w/w) in methanol, solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)4N(OH)
CAS Number:
Molecular Weight:
91.15
Beilstein:
3558708
MDL number:
UNSPSC Code:
12161900
Recommended Products
form
solution
concentration
25 % (w/w) in methanol
storage temp.
2-8°C
SMILES string
[OH-].C[N+](C)(C)C
InChI
1S/C4H12N.H2O/c1-5(2,3)4;/h1-4H3;1H2/q+1;/p-1
InChI key
WGTYBPLFGIVFAS-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Qin Chen et al.
Bioresource technology, 117, 186-192 (2012-05-23)
The aim of this study was to explore the synergies of laccase, a ligninolytic enzyme, with cellulose and hemicellulase amendments on ensiled corn stover. Molecular signals of lignin decomposition were observed by tetramethylammonium hydroxide thermochemolysis and gas chromatography-mass spectroscopy (TMAH-GC-MS)
Hui Chong et al.
Langmuir : the ACS journal of surfaces and colloids, 28(4), 2091-2098 (2011-11-08)
A new water-soluble conjugated polymer containing fluorene and boron-dipyrromethene repeat units in the backbones (PBF) that exhibits red emission was synthesized and characterized. Cationic PBF forms uniform nanoparticles with negatively charged disodium salt 3,3'-dithiodipropionic acid (SDPA) in aqueous solution through
Lina Barreto et al.
Eukaryotic cell, 10(9), 1241-1250 (2011-07-05)
Potassium homeostasis is crucial for living cells. In the yeast Saccharomyces cerevisiae, the uptake of potassium is driven by the electrochemical gradient generated by the Pma1 H(+)-ATPase, and this process represents a major consumer of the gradient. We considered that
Mei Fang et al.
Nanotechnology, 23(14), 145601-145601 (2012-03-22)
Magnetite nanoparticles have been prepared by co-precipitation using a custom-designed jet mixer to achieve rapid mixing (RM) of reactants in a timescale of milliseconds. The quick and stable nucleation obtained allows control of the particle size and size distribution via
Man Xu et al.
Environmental science & technology, 47(1), 110-118 (2012-06-12)
Elucidating dissolution kinetics and mechanisms at carbonate mineral-water interfaces is essential to many environmental and geochemical processes, including geologic CO(2) sequestration in deep aquifers. In the present work, effects of background electrolytes on dolomite (CaMg(CO(3))(2)) reactivity were investigated by measuring
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service