Skip to Content
Merck
All Photos(1)

Documents

SRP6477

Sigma-Aldrich

LDLR human

recombinant, expressed in HEK 293 cells, ≥90% (SDS-PAGE)

Synonym(s):

FH, FHC, LDLCQ2, Low-Density Lipoprotein (LDL) Receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥90% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 86 kDa
observed mol wt 125-140 kDa
observed mol wt 95-110 kDa by SDS-PAGE (Ala22 or Asp193 is the predicted N-terminal.)

packaging

pkg of 10 and 50 μg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... LDLR(3949)

General description

Low-Density Lipoprotein (LDL) Receptor, also known as LDLR, FH, FHC, LDLCQ2, and is a mosaic protein of ~840 amino acids (after removal of signal peptide) that mediates the endocytosis of cholesterol-rich LDL. It is a cell-surface receptor that recognizes the apoprotein B100 which is embedded in the phospholipid outer layer of LDL particles. The receptor also recognizes the apoE protein found in chylomicron remnants and VLDL remnants (IDL). It belongs to the Low density lipoprotein receptor gene family. LDL receptor complexes are present in clathrin-coated pits (or buds) on the cell surface, which when bound to LDL-cholesterol via adaptin, are pinched off to form clathrin-coated vesicles inside the cell. This allows LDL-cholesterol to be bound and internalized in a process known as endocytosis and prevents the LDL just diffusing around the membrane surface. This occurs in all nucleated cells (not erythrocytes), but mainly in the liver which removes ~70% of LDL from the circulation. Synthesis of receptors in the cell is regulated by the level of free intracellular cholesterol; if it is in excess for the needs of the cell then the transcription of the receptor gene will be inhibited. LDL receptors are translated by ribosomes on the endoplasmic reticulum and are modified by the Golgi apparatus before travelling in vesicles to the cell surface. LDL is directly involved in the development of atherosclerosis, due to accumulation of LDL-cholesterol in the blood. Atherosclerosis is the process responsible for the majority of cardiovascular diseases.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergio Fazio et al.
The Journal of clinical endocrinology and metabolism, 102(9), 3340-3348 (2017-06-22)
Angiopoietin-like 3 (ANGPTL3) deficiency in plasma due to loss-of-function gene mutations results in familial combined hypobetalipoproteinemia type 2 (FHBL2) in homozygotes. However, the lipid phenotype in heterozygotes is much milder and does not appear to relate directly to ANGPTL3 levels.
Dalia A Bashir et al.
Clinical and applied thrombosis/hemostasis : official journal of the International Academy of Clinical and Applied Thrombosis/Hemostasis, 27, 1076029621992128-1076029621992128 (2021-02-05)
Hyperferritinemia is associated with poor outcomes in critically ill patients with sepsis, hemophagocytic lymphohistiocytosis (HLH), macrophage activation syndromes (MAS) and coronavirus disease 19 (COVID-19). Autopsies of hyperferritinemic patients that succumbed to either sepsis, HLH, MAS or COVID-19 have revealed disseminated
Altar M Munis et al.
Journal of virology, 92(23) (2018-09-21)
Vesicular stomatitis virus Indiana strain G protein (VSVind.G) is the most commonly used envelope glycoprotein to pseudotype lentiviral vectors (LV) for experimental and clinical applications. Recently, G proteins derived from other vesiculoviruses (VesG), for example, Cocal virus, have been proposed
Yingxue Cui et al.
Acupuncture in medicine : journal of the British Medical Acupuncture Society, 37(4), 237-243 (2019-05-30)
To investigate the anti-atherogenic effect of moxibustion and whether it is mediated through the reverse cholesterol transport process. 8-week-old male apolipoprotein E deficient (ApoE-/- knockout) mice were randomly divided into two groups (n=10 per group): atherosclerosis (AS) and AS plus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service