Skip to Content
Merck
All Photos(1)

Key Documents

SRP6318

Sigma-Aldrich

Kininogen,HMW from human plasma

≥95% (SDS-PAGE)

Synonym(s):

Alpha-2-thiol proteinase inhibitor, BDK, Fitzgerald factor, High molecular weight Kininogen, KNG, Kininogen-1 heavy chain, Kininogen-1 light chain, Low molecular weight growth-promoting, Lysyl-bradykinin, T-kinin Bradykinin, Williams-Fitzgerald-Flaujeac factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human plasma

Assay

≥95% (SDS-PAGE)

form

frozen liquid

mol wt

120 kDa

packaging

pkg of 100 μg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KNG1(3827)

General description

HMW kininogen, synthesized by hepatocytes, is a multifunctional protein. It is the precursor protein of bradykinin. In addition it is a cofactor in the activation of factor XI, XII, and prekallikrein and a major extra-cellular cysteine proteinase inhibitor. Kininogens inhibit systemic proteases which are thought to have a part in various diseases such as cancer, muscular dystrophy and joint disease.

Physical form

Frozen in 10 mM Na acetate, pH 5.5 with 250 mM NaCI.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service