Skip to Content
Merck
All Photos(1)

Key Documents

SRP6251

Sigma-Aldrich

CCL6 from mouse

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

C10, CCL6, MRP-1, SCYA6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

mouse

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

calculated mol wt 11.4 kDa (The predicted N-terminus is Gly22.)
observed mol wt 14 kDa by SDS-PAGE (reducing)

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... CCL6(20305)

General description

Chemokine (C-C motif) ligand 6 (CCL6), a member of the CC chemokine family,[1] was initially identified as a transcript that is induced in bone marrow cells upon stimulation with GM-CSF. CCL6 has only been identified in rodents. A human homologue for mouse CCL6 has not been identified. The mCCL6 cDNA encodes a 116 amino acid residue precursor protein with a hydrophobic signal peptide that is cleaved to yield a 95 amino acid residue mature protein. In mice, CCL6 is expressed in cells from neutrophil and macrophage lineages,[1] where it is strongly induced upon GM-CSF stimulation. Mouse CCL6 expression was also shown to be strongly induced by interleukin-4 (IL-4) in mouse macrophages and by cytokine IL-13 in mouse lung.[2] Mouse CCL6 is located on chromosome 11.[3] The cell surface receptor for CCL6 is believed to be the chemokine receptor CCR1.[2]

Physical form

Lyophilized from 0.22 μm filtered solution in PBS. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The C10/CCL6 chemokine and CCR1 play critical roles in the pathogenesis of IL-13-induced inflammation and remodeling.
Ma B
Journal of Immunology (2004)
The gene for C10, a member of the beta-chemokine family, is located on mouse chromosome 11 and contains a novel second exon not found in other chemokines.
Berger MS
Dna and Cell Biology (1993)
Novel expression pattern of a new member of the MIP-1 family of cytokine-like genes.
Orlofsky A
Cell Regulation (1991)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service