Skip to Content
Merck
All Photos(1)

Key Documents

SRP5360

Sigma-Aldrich

ALK7, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

ACVR1C, ACVRLK7

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
CHF 275.00

CHF 275.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 μG
CHF 275.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

CHF 275.00


Check Cart for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~68 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ACVR1C(130399)

General description

ALK7 is a type I receptor for the TGFB family of signaling molecules in which type I receptors phosphorylate cytoplasmic SMAD transcription factors, which then translocate to the nucleus and interact directly with DNA or in complex with other transcription factors. Alk7 was expressed in all insulin, glucagon, and somatostatin -positive cells of the pancreas. GDF3 regulates adipose tissue homeostasis and energy balance under nutrient overload, in part, by signaling through ALK7 and Alk7 showed reduced fat accumulation and partial resistance to diet-induced obesity, similar to Gdf3.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service