Skip to Content
Merck
All Photos(1)

Key Documents

SRP5358

Sigma-Aldrich

TSSK3, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

SPOGA3, STK22C, STK22D, TSK3

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
CHF 327.00

CHF 327.00


Check Cart for Availability


Select a Size

Change View
20 μG
CHF 327.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

CHF 327.00


Check Cart for Availability

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~56 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TSSK3(81629)

General description

TSSK3 is a serine/threonine kinase belonging to the subfamily of testis-specific kinases 3 which expressed exclusively in the testis that is thought to play a main role in either germ cell differentiation or mature sperm function.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service