Skip to Content
Merck
All Photos(1)

Key Documents

SRP5351

Sigma-Aldrich

AATK, GST tagged from mouse

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

AATYK, KIAA0641, LMTK1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
CHF 328.00

CHF 328.00


Estimated to ship on16 April 2025



Select a Size

Change View
20 μG
CHF 328.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

CHF 328.00


Estimated to ship on16 April 2025


recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~71 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... Aatk(11302)

General description

AATK or apoptosis-associated tyrosine kinase contains a tyrosine kinase domain at the N-terminus and a proline-rich domain at the C-terminus. AATK is induced during apoptosis, and expression of this protein is a necessary pre-requisite for the induction of growth arrest and/or apoptosis of myeloid precursor cells. AATK is highly detected in brain, lung, kidney, and pancreas. AATK is also shown to produce neuronal differentiation in a neuroblastoma cell line.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service