Skip to Content
Merck
All Photos(1)

Key Documents

SRP5345

Sigma-Aldrich

CyclinE1, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CCNE, CCNE1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
CHF 328.00

CHF 328.00


Check Cart for Availability


Select a Size

Change View
20 μG
CHF 328.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

CHF 328.00


Check Cart for Availability

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~73 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CCNE1(898)

General description

Cyclin E1 belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle that functions as regulators of CDK kinases. Cyclin E1 activity is required for centrosome duplication during S phase and this mechanism could coordinate centrosome reproduction with cycles of DNA synthesis and mitosis. The downregulation of cyclin E-CDK2 kinase activity following the G1/S-phase transition that is necessary for the maintenance of karyotypic stability. Cyclin E also has a modular centrosomal-targeting domain which is essential for promoting S phase entry in a Cdk2-independent manner.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dandan Song et al.
Oncology reports, 40(3), 1348-1358 (2018-07-18)
Moxidectin (MOX), a broad‑spectrum antiparasitic agent, belongs to the milbemycin family and is similar to avermectins in terms of its chemical structure. Previous research has revealed that milbemycins, including MOX, may potentially function as effective multidrug resistance agents. In the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service