Skip to Content
Merck
All Photos(2)

Key Documents

SRP5319

Sigma-Aldrich

PI3K (p110-α/p55-γ), active, His tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

p110-alpha, p55-gamma

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CHF 350.00

CHF 350.00


Estimated to ship on11 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
CHF 350.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

CHF 350.00


Estimated to ship on11 May 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

11.2-16.8 nmol/min·mg

mol wt

~111 kDa (p110α)
~61 kDa (p85γ)

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PIK3CA(5290)

General description

PI3K (p110alpha/p55gamma) or Phosphatidylinositol 3-kinase is a lipid kinase that phosphorylates the inositol ring of phosphatidylinositol at the 3-prime position which serve as second messengers in growth signaling pathways. PI3K comprises a 110 kD catalytic subunit and a regulatory subunit of either 85, 55, or 50 kD. Phosphatidylinositol 3-kinase plays an important role in the metabolic actions of insulin, and a mutation in the PI3K has been associated with insulin resistance and plays an important role in glucose homeostasis in vivo. PI3K also plays an essential role in the development and induction of mast cells in normal and pathogenic immune responses.

Physical form

Supplied in 50 mM sodium phosphate, pH 7.0, 300 mM NaCl, 150 mM imidazole, 0.1 mM PMSF, 0.2 mM DTT, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service