Skip to Content
Merck
All Photos(1)

Key Documents

SRP5131

Sigma-Aldrich

SMAD2, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

JV18, JV18-1, MADH2, MADR2, MGC22139, MGC34440, hMAD-2, hSMAD2

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
CHF 286.00

CHF 286.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
CHF 286.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

CHF 286.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~90 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SMAD2(4087)

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Targeting the transforming growth factor-beta signaling pathway in human cancer.
Nagaraj NS
Expert Opinion on Investigational Drugs (2010)
Signaling of transforming growth factor-beta family members through Smad proteins.
Itoh S
European Journal of Biochemistry (2000)
Alterations in the Smad pathway in human cancers.
Samanta D and Datta PK
Frontiers in Bioscience (Landmark Edition) (2012)
K Eppert et al.
Cell, 86(4), 543-552 (1996-08-23)
The MAD-related (MADR) family of proteins are essential components in the signaling pathways of serine/threonine kinase receptors for the transforming growth factor beta (TGFbeta) superfamily. We demonstrate that MADR2 is specifically regulated by TGFbeta and not bone morphogenetic proteins. The
M Funaba et al.
Molecular endocrinology (Baltimore, Md.), 14(10), 1583-1591 (2000-10-24)
Smads mediate activin, transforming growth factor beta (TGFbeta), and bone morphogenetic protein signaling from receptors to nuclei. According to the current model, activated activin/TGFbeta receptors phosphorylate the carboxyl-terminal serines of Smad2 and Smad3 (SSMS-COOH); phosphorylated Smad2/3 oligomerizes with Smad4, translocates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service