Skip to Content
Merck
All Photos(1)

Key Documents

SRP5105

Sigma-Aldrich

NFKB2 (1-454), GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

LYT-10, LYT10, NFKB2-p100, NFKB2-p52

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~84 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NFKB2(4791)

General description

NFKB2 is a part of the NFKB complex that is present in various cell types that express cytokines, chemokines, growth factors, cell adhesion molecules, and some acute phase proteins in health and disease states. NFKB protein is activated by a wide variety of stimuli such as cytokines, oxidant-free radicals, inhaled particles, ultraviolet irradiation, and bacterial or viral products. Inappropriate activation of NFKB has been linked to inflammatory events associated with autoimmune arthritis, asthma, septic shock, lung fibrosis, glomerulonephritis, atherosclerosis, and AIDS. Overexpression of NFKB2 protects androgen sensitive LNCaP cells from apoptotic cell death and cell cycle arrest induced by androgen-deprivation.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena Tucker et al.
Journal of immunology (Baltimore, Md. : 1950), 179(11), 7514-7522 (2007-11-21)
The noncanonical NF-kappaB pathway regulates the development and function of multiple organs and cell lineages. We have generated mice harboring a novel mutation in Nfkb2 that prevents the processing of the inhibitory precursor, p100, into the active subunit, p52. Mutant
Mingzhao Zhu et al.
The Journal of clinical investigation, 116(11), 2964-2971 (2006-10-14)
NF-kappaB2-deficient mice have impaired T and B cell responses. We found, however, that in these mice there was severe infiltration of lymphocytes into multiple organs and increased activity of autoantibodies to peripheral tissue antigens in a manner similar to that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service