SRP5069
PRKG2, active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
PRKGR2, cGKII
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
recombinant
expressed in baculovirus infected Sf9 cells
product line
PRECISIO® Kinase
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
228-308 nmol/min·mg
mol wt
~112 kDa
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... PRKG2(5593)
General description
PRKG2 is a member of the cGMP-dependent protein kinase family and is highly expressed in brain, lung, and intestinal mucosa. PRKG2 mediates intestinal secretion of water and electrolytes induced by the E. coli toxin STa and the intestinal peptide guanylin. Mice deficient in PRKG2 are resistant to E. coli STa and developed dwarfism which is caused by a severe defect in endochondral ossification at the growth plates. PRKG2 phosphorylates SOX9 and attenuates SOX9 function by inhibiting its nuclear entry. PRKG2 is a molecular switch that couples the cessation of proliferation and the start of hypertrophic chondrocyte differentiation through attenuating SOX9 function.
Physical form
Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.
Preparation Note
after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Genes & development, 18(19), 2418-2429 (2004-10-07)
The Komeda miniature rat Ishikawa (KMI) is a naturally occurring mutant caused by an autosomal recessive mutation mri, which exhibits longitudinal growth retardation. Here we identified the mri mutation as a deletion in the rat gene encoding cGMP-dependent protein kinase
Science (New York, N.Y.), 274(5295), 2082-2086 (1996-12-20)
Cyclic guanosine 3',5'-monophosphate (cGMP)-dependent protein kinases (cGKs) mediate cellular signaling induced by nitric oxide and cGMP. Mice deficient in the type II cGK were resistant to Escherichia coli STa, an enterotoxin that stimulates cGMP accumulation and intestinal fluid secretion. The
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service