Skip to Content
Merck
All Photos(1)

Key Documents

SRP3245

Sigma-Aldrich

IL-9 from rat

≥98% (SDS-PAGE & HPLC), recombinant, expressed in E. coli, lyophilized, suitable for cell culture

Synonym(s):

T-cell growth factor p40, p40 cytokine

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CHF 333.00

CHF 333.00


Estimated to ship on04 May 2025



Select a Size

Change View
10 μG
CHF 333.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

CHF 333.00


Estimated to ship on04 May 2025


Product Name

IL-9 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

biological source

rat

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

14.3 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... IL9(116558)

General description

IL-9 is an immunoregulatory cytokine produced by IL-2 activated Th2 lymphocytes. IL-9 enhances the proliferation of T lymphocytes, mast cells, erthroid precursor cells and megakaryoblastic leukemia cell lines. Over-expression of IL-9 has been implicated in the pathogenesis of anaplastic lymphoma and Hodgkin′s disease. Recombinant rat IL-9 is a 14.3 kDa protein of 127 amino acid residues including 10 cysteine residues which are fully conserved between the human and murine proteins.

Biochem/physiol Actions

IL-9 is an immunoregulatory cytokine produced by IL-2 activated Th2 lymphocytes. Recombinant rat IL-9 is a 14.3 kDa protein of 127 amino acid residues including 10 cysteine residues which are fully conserved between the human and murine proteins.

Sequence

MQRCSTSWGI QHTSYLIENL KDDPSSKCSC SANVTSCLCL PIPSDDCTTP CFQEGMSQVT NATQQSKFSP FFFRVKRIVE TLKSNKCQFF SCEKPCNQTT AGNTVSFLKS LLKTFQKTEV QVQRSRA

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rollins, R.J., et al.
Cytokine Reference, 1145-1160 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service