Skip to Content
Merck
All Photos(1)

Key Documents

SRP3218

Sigma-Aldrich

LIX (70 aa) from mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CXCL5

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
CHF 347.00

CHF 347.00


Check Cart for Availability


Select a Size

Change View
20 μG
CHF 347.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

CHF 347.00


Check Cart for Availability

biological source

mouse

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

7.6 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... CXCL5(20311)

General description

The murine homolog of ENA-78 is called LIX. LIX is a CXC chemokine that signals through the CXCR2 receptor. It is expressed in monocytes, platelets, endothelial cells, and mast cells. LIX is a chemoattractant for neutrophils. The two naturally occurring variants of LIX; LIX 1-78 (GCP-2) and LIX 9-78 (GCP-2), contain 78 and 70 amino acid residues, respectively. LIX contains the four conserved cysteine residues present in CXC chemokines, and also contains the ‘ELR′ motif common to CXC chemokine that bind to the CXCR1 and CXCR2 receptors. Recombinant murine LIX is a 7.6 kDa protein consisting of amino acid residues 9-78 of the full length LIX protein (70 amino acids).

Biochem/physiol Actions

The murine homolog of ENA-78 is called LIX. LIX is a CXC chemokine that signals through the CXCR2 receptor. Recombinant murine LIX is a 7.6 kDa protein consisting of amino acid residues 9-78 of the full length LIX protein (70 amino acids).

Sequence

TELRCVCLTV TPKINPKLIA NLEVIPAGPQ CPTVEVIAKL KNQKEVCLDP EAPVIKKIIQ KILGSDKKKA

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N W Lukacs et al.
Journal of leukocyte biology, 63(6), 746-751 (1998-06-10)
The inflammatory response during allergic airway inflammation involves the recruitment of multiple leukocyte populations, including neutrophils, monocytes, lymphocytes, and eosinophils. All of these populations likely contribute to the pathology observed during repeated episodes of allergic airway inflammation. We have examined

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service