SRP3123
Neuroserpin human
recombinant, expressed in E. coli, ≥96% (SDS-PAGE), ≥96% (HPLC), suitable for cell culture
Synonym(s):
Protease inhibitor 12, Serpin I1
Select a Size
CHF 319.00
Select a Size
About This Item
CHF 319.00
Recommended Products
biological source
human
recombinant
expressed in E. coli
Assay
≥96% (HPLC)
≥96% (SDS-PAGE)
form
lyophilized
mol wt
44.6 kDa
packaging
pkg of 25 μg
technique(s)
cell culture | mammalian: suitable
impurities
<0.1 EU/μg endotoxin, tested
color
white to off-white
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... SERPINI1(5274)
General description
Biochem/physiol Actions
Sequence
Physical form
Reconstitution
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service