Skip to Content
Merck
All Photos(1)

Key Documents

SRP2101

Sigma-Aldrich

Myc-associated factor X (MAX) human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

MGC10775, MGC11225, MGC18164, MGC34679, bHLHd4, bHLHd5, bHLHd6, bHLHd7, bHLHd8

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CHF 466.00

CHF 466.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
10 μG
CHF 466.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

CHF 466.00


Estimated to ship on16 April 2025


Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~19.6 kDa

packaging

pkg of 10 μg

concentration

250 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAX(4149)

General description

The gene encoding this protein is localized on human chromosome 14q23.[1]

Biochem/physiol Actions

The MAX gene encodes a protein that interacts specifically with the Myc protein to form a heterodimer with high affinity for the specific cognate DNA-binding site of Myc. The protein encoded by this gene is a member of the basic helix-loop-helix leucine zipper (bHLHZ) family of transcription factors. It is able to form homodimers and heterodimers with other family members, which include MAD, MXI1 and Myc. Myc is an oncoprotein implicated in cell proliferation, differentiation and apoptosis. The homodimers and heterodimers compete for a common DNA target site (the E box) and rearrangement among these dimer forms provides a complex system of transcriptional regulation. Substantial evidence has been accumulated over the last years that support the model that Myc/MAX/MAD proteins affect different aspects of cell behavior, including proliferation, differentiation, and apoptosis, by modulating distinct target genes. The unbalanced expression of these genes, e.g. in response to deregulated Myc expression, is most likely an important aspect of Myc`s ability to stimulate tumor formation. It is reported that in vivo transactivation assays suggest that Myc-MAX and MAD-MAX complexes have opposing functions in transcription and that MAX plays a central role in this network of transcription factors. High levels of MAX and stress-induced NFkappaB activation may result in elevated expression of Fas ligand in human lung cancer cells and possibly contribute to Fas ligand-associated immune escape mechanisms.
This protein has been associated with familial pheochromocytoma (PCC).[2][3]

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hypoxia reduces MAX expression in endothelial cells by unproductive splicing
Katrin Kemmerer
Febs Letters, 588 (2014)
Complex MAX Rearrangement in a Family With Malignant Pheochromocytoma, Renal Oncocytoma, and Erythrocytosis
Esther Korpershoek
The Journal of Clinical Endocrinology and Metabolism, 101(2), 453-460 (2016)
B Lüscher
Gene, 277(1-2), 1-14 (2001-10-17)
The members of the Myc/Max/Mad network function as transcriptional regulators. Substantial evidence has been accumulated over the last years that support the model that Myc/Max/Mad proteins affect different aspects of cell behavior, including proliferation, differentiation, and apoptosis, by modulating distinct
E M Blackwood et al.
Science (New York, N.Y.), 251(4998), 1211-1217 (1991-03-08)
The myc protooncogene family has been implicated in cell proliferation, differentiation, and neoplasia, but its mechanism of function at the molecular level is unknown. The carboxyl terminus of Myc family proteins contains a basic region helix-loop-helix leucine zipper motif (bHLH-Zip)
Zoltan Wiener et al.
Experimental cell research, 299(1), 227-235 (2004-08-11)
Fas (CD95/APO-1) ligand is a member of the Tumor Necrosis Factor family and a potent inducer of apoptosis. Fas ligand is expressed in activated T cells and represents a major cytotoxic effector mechanism by which T cells kill their target

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service