Skip to Content
Merck
All Photos(1)

Key Documents

SRP2079

Sigma-Aldrich

p300 human

recombinant, expressed in insect cells, ≥70% (SDS-PAGE)

Synonym(s):

KAT3B, RP1-85F18.1, RSTS2, p300

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

biological source

human

recombinant

expressed in insect cells

Assay

≥70% (SDS-PAGE)

form

frozen liquid

mol wt

~265 kDa

packaging

pkg of 4 μg

concentration

500 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... EP300(2033)

General description

E1A binding protein p300 (p300) has a central bromodomain and three cysteine- and histidine-rich regions. The carboxy-terminal region interacts with the adenovirus oncoprotein E1A. The gene encoding this protein is localized on human chromosome 22q13. P300 is full length recombinant protein with flag tag at its N-terminus.

Application

p300 human has been used in in vitro acetylation assay and in vitro kinase assay of mechanistic target of rapamycin complex 1 (mTORC1).

Biochem/physiol Actions

EP300 and CREBBP are highly related transcriptional coactivators mediating cAMP gene regulation. EP300 is known to acetylate many transcription factors, including p53, E2F, TFIIE, and TFIIF.
EP300 and CREBBP are highly related transcriptional coactivators mediating cAMP gene regulation. EP300 is known to acetylate many transcription factors, including p53, E2F, TFIIE, and TFIIF.

p300 activates transcription of E-cadherin, which helps to retain the cell in an epithelial state. p300 plays an essential role in integrating and mediating multiple signal-dependent transcription events. It acts as a histone acetyltransferase and controls transcription through chromatin remodeling. Mutation in the gene has been observed in various types of malignancies.
Human p300 and CBP (CREB binding protein) are highly related transcriptional coactivators. Both proteins have been identified through protein interaction assays. In addition to interacting with a variety of cellular factors and onco-proteins, loss of the wild type CBP alleles in isolated tumors suggests that CBP/p300 might serve as tumor suppressors. The ability of p300 to acetylate many transcription factors, including p53, E2F, TFIIE, and TFIIF etc. demonstrates a novel mechanism of targeted p300 regulation of gene expression.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

mTORC1 Phosphorylates Acetyltransferase p300 to Regulate Autophagy and Lipogenesis.
Wan W, et al.
Molecular Cell, 68(2), 323-335 (2017)
Pedro Latorre-Muro et al.
The Journal of biological chemistry, 296, 100205-100205 (2020-12-19)
Acetylation is known to regulate the activity of cytosolic phosphoenolpyruvate carboxykinase (PCK1), a key enzyme in gluconeogenesis, by promoting the reverse reaction of the enzyme (converting phosphoenolpyruvate to oxaloacetate). It is also known that the histone acetyltransferase p300 can induce
Acetylation-dependent regulation of MDM2 E3 ligase activity dictates its oncogenic function.
Nihira N T, et al.
Science Signaling, 10(466), eaai8026-eaai8026 (2017)
Tumour suppressor EP300, a modulator of paclitaxel resistance and stemness, is downregulated in metaplastic breast cancer.
Asaduzzaman M, et al.
Breast Cancer Research and Treatment, 163(3), 461-474 (2017)
R W Stein et al.
Journal of virology, 64(9), 4421-4427 (1990-09-01)
Adenovirus E1A transforming function requires two distinct regions of the protein. Transforming activity is closely linked with the presence of a region designated conserved domain 2 and the ability of this region to bind the product of the cellular retinoblastoma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service