Skip to Content
Merck
All Photos(1)

Key Documents

SRP2036

Sigma-Aldrich

BRG1, wild type human

recombinant, expressed in insect cells, ≥80% (SDS-PAGE)

Synonym(s):

BAF190, BRG1, FLJ39786, RTPS2, SNF2, SNF2-beta, SNF2L4, SNF2LB, SWI2, hSNF2b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

biological source

human

recombinant

expressed in insect cells

Assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~185 kDa

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

300 μg/mL

color

clear
colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SMARCA4(6597)

Biochem/physiol Actions

The wild type human brahma-related gene 1 (Brg1) encodes a protein of 1,647 amino acids that contains a conserved domain of the SWI2/SNF2 family necessary for normal mitotic growth and transcription regulation. BRG1 is an essential component of the SWI/SNF chromatin remodeling complexes and implicated in multiple functions through its interaction with different proteins, including the tumor suppressor protein pRb, serine-threonine kinase LKB1, and other transcription factors. Although Brg1 is involved in chromatin remodeling as a complex with other SWI/SNF proteins, purified BRG1 itself is capable of remodeling mono-nucleosomes and nucleosomal arrays in vitro. Mutations of Brg1 have been found in multiple tumor cell lines.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Sif et al.
Genes & development, 12(18), 2842-2851 (1998-09-23)
During mitosis, chromatin is condensed into mitotic chromosomes and transcription is inhibited, processes that might be opposed by the chromatin remodeling activity of the SWI/SNF complexes. Brg1 and hBrm, which are components of human SWI/SNF (hSWI/SNF) complexes, were recently shown
C J Fryer et al.
Nature, 393(6680), 88-91 (1998-05-20)
The assembly of transcriptional regulatory DNA sequences into chromatin plays a fundamental role in modulating gene expression. The promoter of the mouse mammary-tumour virus (MMTV) is packaged into a regular array of nucleosomes when it becomes stably integrated into mammalian
P A Khavari et al.
Nature, 366(6451), 170-174 (1993-11-11)
Sequence-specific DNA binding activators of gene transcription may be assisted by SWI2 (SNF2), which contains a DNA-dependent ATPase domain. We have isolated a human complementary DNA encoding a 205K nuclear protein, BRG1, that contains extensive homology to SWI2 and Drosophila

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service