Skip to Content
Merck
All Photos(1)

Key Documents

SRP0556

Sigma-Aldrich

Human CD137 (4-1BB), FC Fusion

recombinant, expressed in HEK 293 cells

Synonym(s):

ILA, TNFRSF9, Tumor Necrosis Factor Receptor Superfamily member 9

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 545.00

CHF 545.00


Estimated to ship on20 May 2025



Select a Size

Change View
100 μG
CHF 545.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

CHF 545.00


Estimated to ship on20 May 2025


recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

44 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TNFRSF9(3604)

General description

Human secreted CD137, Fc fusion protein, also known as Tumor Necrosis Factor Receptor Superfamily member 9, TNFRSF9, and ILA, GenBank Accession No. NM_001561, a.a. 24-186 expressed in a HEK293 cell expression system. MW = 44 kDa (monomer). This protein runs at a higher MW by SDS-PAGE due to glycosylation.

Physical form

Aqueous solution

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service