Skip to Content
Merck
All Photos(1)

Key Documents

SRP0485

Sigma-Aldrich

Bromodomain Ligand 2

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 ML
CHF 670.00

CHF 670.00


Check Cart for Availability


Select a Size

Change View
0.5 ML
CHF 670.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

CHF 670.00


Check Cart for Availability

Assay

≥95% (HPLC)

form

aqueous solution

mol wt

2765.3 kDa

packaging

pkg of 5 mL

shipped in

dry ice

storage temp.

−20°C

General description

The Bromodomain Ligand 2 is peptide histone H3 (1-21), acetylated at lysine 14. It contains a C-terminal GG linker, followed by a biotinylated Lys. Bromodomains function as acetyl-lysine binding domains, and bromodomain/acetyl-lysine recognition can regulate histone modification, DNA replications, chromatin remodeling, and gene transcription. This peptide is suitable for use as a ligand for several bromodomains including ATAD2 and BAZ2B.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C A Johnson et al.
Nucleic acids research, 26(4), 994-1001 (1998-03-21)
The pattern of histone H4 acetylation in different genomic regions has been investigated by immunoprecipitating oligonucleosomes from a human lymphoblastoid cell line with antibodies to H4 acetylated at lysines 5, 8, 12 or 16. DNA from antibody-bound or unbound chromatin
Z Jasencakova et al.
The Plant cell, 12(11), 2087-2100 (2000-11-23)
Reversible acetylation of nucleosomal histones H3 and H4 generally is believed to be correlated with potential transcriptional activity of eukaryotic chromatin domains. Here, we report that the extent of H4 acetylation within euchromatin and heterochromatic domains is linked with DNA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service