Skip to Content
Merck
All Photos(1)

Key Documents

SRP0482

Sigma-Aldrich

ATAD2B (953-1080) human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

ATPase family, AAA domain containing 2B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

15.9 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ATAD2B(54454)

General description

Human bromodomain ATPase family, AAA domain containing 2B (variant 1) (GenBank Accession No. NM_017552) amino-acids 951 - 1080 with N-terminal HIS-tag, MW = 15.9 kDa, expressed in an E. coli expression system.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathaniel T Leachman et al.
Development, growth & differentiation, 52(9), 747-755 (2010-12-17)
ATAD2 is an E2F target gene that is highly expressed in gastrointestinal and breast carcinomas. Here we characterize a related gene product, ATAD2B. Both genes are evolutionarily conserved, with orthologues present in all eukaryotic genomes examined. Human ATAD2B shows a
Toshio Ota et al.
Nature genetics, 36(1), 40-45 (2004-01-01)
As a base for human transcriptome and functional genomics, we created the "full-length long Japan" (FLJ) collection of sequenced human cDNAs. We determined the entire sequence of 21,243 selected clones and found that 14,490 cDNAs (10,897 clusters) were unique to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service