Skip to Content
Merck
All Photos(1)

Key Documents

SRP0475

Sigma-Aldrich

TAF1 (1519-1651) human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

DYT3, KAT4, TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 250 kDa, TAF2ADYT3/TAF1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 670.00

CHF 670.00


Estimated to ship on19 April 2025



Select a Size

Change View
100 μG
CHF 670.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

CHF 670.00


Estimated to ship on19 April 2025


biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

16.4 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TAF1(6872)

General description

Human Bromodomain TAF1 (GenBank Accession No. NM_004606) amino acids 1519 – 1651 with N-terminal HIS-tag, MW = 16.4 kDa, expressed in an E. coli expression system.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Sako et al.
Neuroscience, 189, 100-107 (2011-05-28)
The neuron-specific isoform of the TAF1 gene (N-TAF1) is thought to be involved in the pathogenesis of DYT3 dystonia, which leads to progressive neurodegeneration in the striatum. To determine the expression pattern of N-TAF1 transcripts, we developed a specific monoclonal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service