SRP0449
BRD1 (561-668) GST tag human
recombinant, expressed in E. coli, ≥90% (SDS-PAGE)
Synonym(s):
BRL, Bromodomain containing 1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
Recommended Products
biological source
human
recombinant
expressed in E. coli
Assay
≥90% (SDS-PAGE)
form
aqueous solution
mol wt
40 kDa
packaging
pkg of 100 μg
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... BRD1(23774)
General description
Human Bromodomain containing 1, also known as BRD1 and BRL (GenBank Accession No. NM_014577) amino-acids 561 – 668 with N-terminal GST-tag, MW = 40 kDa, expressed in an E. coli expression system.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J E Severinsen et al.
Molecular psychiatry, 11(12), 1126-1138 (2006-08-23)
Linkage studies suggest that chromosome 22q12-13 may contain one or more shared susceptibility genes for schizophrenia (SZ) and bipolar affective disorder (BPD). In a Faeroese sample, we previously reported association between microsatellite markers located at 22q13.31-qtel and both disorders. The
Yuta Mishima et al.
Blood, 118(9), 2443-2453 (2011-07-15)
The histone acetyltransferases (HATs) of the MYST family include TIP60, HBO1, MOZ/MORF, and MOF and function in multisubunit protein complexes. Bromodomain-containing protein 1 (BRD1), also known as BRPF2, has been considered a subunit of the MOZ/MORF H3 HAT complex based
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service