Skip to Content
Merck
All Photos(2)

Key Documents

SRP0442

Sigma-Aldrich

UEV1A active human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

CIR1, CROC1, UBE2V1, ubiquitin-conjugating enzyme E2 variant 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 513.00

CHF 513.00


Estimated to ship on29 April 2025



Select a Size

Change View
100 μG
CHF 513.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

CHF 513.00


Estimated to ship on29 April 2025


biological source

human

recombinant

expressed in E. coli

Assay

≥80% (SDS-PAGE)

form

aqueous solution

mol wt

20 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... UBE2V1(7335)

General description

UEV1A (ubiquitin-conjugating enzyme E2 variant 1), also known as UBE2V1 or CROC-1 (GenBank Accession No. NM_021988) amino acids 2-170 (end) with N-terminal FLAG-tag, MW=20 kDa, expressed in E. coli.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Ma et al.
Oncogene, 17(10), 1321-1326 (1998-10-15)
Acquisition of the immortal phenotype by tumor cells represents an essential and potentially rate-limiting step in tumorigenesis. To identify changes in gene expression that are associated with the early stages of cell immortalization, we compared genetically matched pairs of pre-immortal
Jing Geng et al.
Nature immunology, 16(11), 1142-1152 (2015-09-29)
Mitochondria need to be juxtaposed to phagosomes for the synergistic production of ample reactive oxygen species (ROS) in phagocytes to kill pathogens. However, how phagosomes transmit signals to recruit mitochondria has remained unclear. Here we found that the kinases Mst1
T M Thomson et al.
FEBS letters, 423(1), 49-52 (1998-05-15)
The open reading frame YGL087c in the budding yeast Saccharomyces cerevisiae genome encodes a polypeptide highly similar to the human UEV (ubiquitin-conjugating E2 enzyme variant) proteins, which have been proposed to belong to a family of putative dominant negative ubiquitin
D Duong Hau et al.
Biochemistry, 45(32), 9866-9877 (2006-08-09)
Lys(63)-linked polyubiquitination of TRAF2 or TRAF6 is an essential step within the signal transduction cascade responsible for activation of p38, c-Jun N-terminal kinase, and the transcription factor NF-kappaB. Attachment of ubiquitin (Ub) to a TRAF, and conjugation of Ub molecules

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service