Skip to Content
Merck
All Photos(1)

Key Documents

SRP0421

Sigma-Aldrich

DPY30 human

recombinant, expressed in E. coli, ≥85% (SDS-PAGE)

Synonym(s):

HDPY-30, Saf19, dpy-30 homolog

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
CHF 670.00

CHF 670.00


Check Cart for Availability


Select a Size

Change View
50 μG
CHF 670.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

CHF 670.00


Check Cart for Availability

biological source

human

recombinant

expressed in E. coli

Assay

≥85% (SDS-PAGE)

form

aqueous solution

mol wt

12 kDa

packaging

pkg of 50 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... DPY30(84661)

General description

Human dpy-30 homolog, or DPY30 (GenBank Accession No. NM_032574) amino acids 2 – 99 (end) with N-terminal His-tag, MW= 12 kDa, expressed in an E. coli expression system.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhuojin Xu et al.
The Journal of cell biology, 186(3), 343-353 (2009-08-05)
Histone lysine methyltransferase complexes are essential for chromatin organization and gene regulation. Whether any of this machinery functions in membrane traffic is unknown. In this study, we report that mammal Dpy-30 (mDpy-30), a subunit of several histone H3 lysine 4
Anamika Patel et al.
The Journal of biological chemistry, 284(36), 24242-24256 (2009-06-27)
Transcription in eukaryotic genomes depends on enzymes that regulate the degree of histone H3 lysine 4 (H3K4) methylation. The mixed lineage leukemia protein-1 (MLL1) is a member of the SET1 family of H3K4 methyltransferases and is frequently rearranged in acute

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service