Skip to Content
Merck
All Photos(1)

Key Documents

SRP0376

Sigma-Aldrich

UTX human

recombinant, expressed in baculovirus infected Sf9 cells, ≥40% (SDS-PAGE)

Synonym(s):

KABUK2, KDM6A, lysine (K)-specific demethylase 6A

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
CHF 670.00

CHF 670.00


Estimated to ship on10 May 2025



Select a Size

Change View
20 μG
CHF 670.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

CHF 670.00


Estimated to ship on10 May 2025


biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥40% (SDS-PAGE)

form

aqueous solution

mol wt

155 kDa

packaging

pkg of 20 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KDM6A(7403)

General description

Human UTX, also known as KDM6A (GenBank Accession No. NM_021140) amino-acids 1-1401 with C-terminal FLAG-tag, MW = 155 kDa, expressed in Sf9 cells via a Baculovirus expression system.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karl Agger et al.
Nature, 449(7163), 731-734 (2007-08-24)
The trithorax and the polycomb group proteins are chromatin modifiers, which play a key role in the epigenetic regulation of development, differentiation and maintenance of cell fates. The polycomb repressive complex 2 (PRC2) mediates transcriptional repression by catalysing the di-
A Greenfield et al.
Human molecular genetics, 7(4), 737-742 (1998-05-16)
We recently have identified a ubiquitously transcribed mouse Y chromosome gene, Uty , which encodes a tetratricopeptide repeat (TPR) protein. A peptide derived from the UTY protein confers H-Y antigenicity on male cells. Here we report the characterization of a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service