Skip to Content
Merck
All Photos(1)

Documents

SRP0286

Sigma-Aldrich

Sphingosine kinase 2 Active from mouse

recombinant, expressed in baculovirus, ≥70% (SDS-PAGE)

Synonym(s):

SK2, SPHK2, SPK 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in baculovirus

Assay

≥70% (SDS-PAGE)

form

aqueous solution

specific activity

≥2 units/μg

mol wt

68 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... SPHK2(56848)

General description

Mouse Sphingosine kinase 2 ( GenBank Accession No. NM_203280), full length with N-terminal His tag, MW = 68 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Sphingosine kinase 2 is useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling. Sphingosine kinase 2 has been used in a study to assess blood–brain barrier protection in hypoxic preconditioning-induced cerebral ischemic tolerance. Sphingosine kinase 2 has also been used in a study to investigate sensitization of human colon cancer cells to sodium butyrate-induced apoptosis.

Unit Definition

One unit of sphingosine kinase activity is defined as the amount of enzyme required to produce 1 pmol of S1P/min

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 50% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Xiao et al.
Experimental cell research, 318(1), 43-52 (2011-10-26)
Sphingosine kinases (SphKs) have been recognized as important proteins regulating cell proliferation and apoptosis. Of the two isoforms of SphK (SphK1 and SphK2), little is known about the functions of SphK2. Sodium butyrate (NaBT) has been established as a promising
Mithun R Raje et al.
Bioorganic & medicinal chemistry, 20(1), 183-194 (2011-12-06)
Sphingosine kinase (SphK) has emerged as an attractive target for cancer therapeutics due to its role in cell survival. SphK phosphorylates sphingosine to form sphingosine 1-phosphate (S1P), which has been implicated in cancer growth and survival. SphK exists as two
Bradley K Wacker et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 32(6), 1014-1023 (2012-02-09)
Protection of the blood-brain barrier (BBB) is correlated with improved outcome in stroke. Sphingosine kinase (SphK)-directed production of sphingosine-1-phosphate, which we previously documented as being vital to preconditioning-induced stroke protection, mediates peripheral vascular integrity via junctional protein regulation. We used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service