Skip to Content
Merck
All Photos(2)

Key Documents

SRP0149

Sigma-Aldrich

Set7/Set9 human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE)

Synonym(s):

KMT7, Lysine N-methyltransferase 7, SET domain containing (lysine methyltransferase) 7, SET7/9, SETD7

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 620.00

CHF 620.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 620.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

CHF 620.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

aqueous solution

mol wt

61 kDa

packaging

pkg of 100 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SETD7(80854)

General description

Human recombinant SET7/9, amino acids 52-end, with N-terminal GST tag, GenBank Accession No. NM_030648, expressed in a E. coli expression system. MW = 61 kDa,
SET7/9 also known as KMT7/SET domain containing lysine methyltransferase 7 (SETD7), is encoded by the gene mapped to human chromosome 4q31.1.[1] The encoded protein belongs to SET domain-containing protein family.[2]

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Biochem/physiol Actions

SET7/9 acts as a histone methyltransferase and catalyzes monomethylation of histone H3 at lysine 4 (H3K4me1) in vitro.[3] Down-regulation of SET7/9 expression affects human embryonic stem cell differentiation.[3] The encoded protein is implicated in the regulation of proteins and transcription factor activity. It also stimulates promoters of methylation-dependent co-recruitment through non-histone protein methylation.[2] Mutation in the gene encoding SET7/9 is associated with serum lycopene concentrations.[1] Elevated expression of the gene is associated with progression of hepatocellular carcinoma (HCC).[2]

Unit Definition

One unit is defined as the amount of enzyme required to methylate 1 pmol of substrate/min at 37°C.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 30% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SETD7 Regulates the Differentiation of Human Embryonic Stem Cells.
Casta?o J
PLoS ONE, 11(2), 1-21 (2016)
A Common Variant in the SETD7 Gene Predicts Serum Lycopene Concentrations.
D'Adamo CR
Nutrients, 8(2):82, 1-11 (2016)
ABCG1 and ABCG4 Suppress ?-Secretase Activity and Amyloid ? Production.
Sano O
PLoS ONE, 11(5), 1-19 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service