Skip to Content
Merck
All Photos(1)

Key Documents

SRP0143

Sigma-Aldrich

PRMT4 Active human

recombinant, expressed in baculovirus infected insect cells, ≥65% (SDS-PAGE)

Synonym(s):

Arginine N-methyltransferase 4, CARM1, Coactivator-associated arginine methyltransferase 1, PRMT4

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
CHF 620.00

CHF 620.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 μG
CHF 620.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

CHF 620.00


Check Cart for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected insect cells

Assay

≥65% (SDS-PAGE)

form

aqueous solution

mol wt

66 kDa

packaging

pkg of 20 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CARM1(10498)

General description

Human PRMT4 (CARM1) (GenBank Accession No. NM_199141), amino acids 2-end, with N-terminal His-tag, MW = 66 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Unit Definition

One unit is defined as the amount of enzyme required to methylate 1 pmol of substrate/min at 37°C.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 30% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatsuo Ito et al.
BMC developmental biology, 9, 47-47 (2009-09-04)
Chondrogenesis and subsequent endochondral ossification are processes tightly regulated by the transcription factor Sox9 (SRY-related high mobility group-Box gene 9), but molecular mechanisms underlying this activity remain unclear. Here we report that coactivator-associated arginine methyltransferase 1 (CARM1) regulates chondrocyte proliferation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service